Product Details
- SNP ID
-
rs3809852
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4439855 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCACAGAGGGTGGGTGGGGTATGG[A/G]CCTGAGGGACAGTCTAGGCAGGAGC
- Phenotype
-
MIM: 611701
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPNS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78337983] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPNS3
- Gene Name
- sphingolipid transporter 3 (putative)
View Full Product Details