Product Details

SNP ID
rs61736206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14054834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCCCTGTCCTCCTTCACCCAGG[C/G]CGGGAGCTCCAGGCTACCAGCTCCC
Phenotype
MIM: 605350
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
There are no transcripts associated with this gene.

Gene
PALM3
Gene Name
paralemmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145028.1 793 Missense Mutation CCC,GCC P265A NP_001138500.1
XM_005259891.4 793 Missense Mutation CCC,GCC P280A XP_005259948.1
XM_011527965.2 793 Missense Mutation CCC,GCC P253A XP_011526267.1

View Full Product Details