Product Details

SNP ID
rs61747644
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:143213279 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAGCGTCCGGCGAGATGAAGGCGCT[C/T]GGGGCTGTCCTGCTTGCCCTCTTGC
Phenotype
MIM: 612757
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPIHBP1 PubMed Links

Gene Details

Gene
GPIHBP1
Gene Name
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301772.1 87 Silent Mutation CTC,CTT L4L NP_001288701.1
NM_178172.5 87 Silent Mutation CTC,CTT L4L NP_835466.2

View Full Product Details