Product Details

SNP ID
rs10792156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58267071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGTGTACACCAGTGAGATGAAC[C/T]GATCTTGCTTGGGGTTGTAGCTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10W1 PubMed Links

Gene Details

Gene
OR10W1
Gene Name
olfactory receptor family 10 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207374.3 1190 Missense Mutation CAG,CGG Q263R NP_997257.2

View Full Product Details