Product Details

SNP ID
rs61750934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36638866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTTACATTCATAGGGTTTCTC[A/G]CCAGTATGAATTCTTTGATGTTGAA
Phenotype
MIM: 608640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF461 PubMed Links

Gene Details

Gene
ZNF461
Gene Name
zinc finger protein 461
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297623.2 1686 Silent Mutation NP_001284552.1
NM_001322821.1 1686 Silent Mutation NP_001309750.1
NM_001322823.1 1686 Silent Mutation NP_001309752.1
NM_001322825.1 1686 Silent Mutation NP_001309754.1
NM_001322826.1 1686 Silent Mutation NP_001309755.1
NM_001322827.1 1686 Silent Mutation NP_001309756.1
NM_001322828.1 1686 Silent Mutation NP_001309757.1
NM_153257.4 1686 Silent Mutation NP_694989.2
XM_011527486.2 1686 Silent Mutation XP_011525788.1
XM_017027470.1 1686 Silent Mutation XP_016882959.1

View Full Product Details