Product Details

SNP ID
rs7229802
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62524195 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGGCTTGGCTCCAGGGCCCAGGA[A/G]GAACTGCTGCTGCTCTTCACCATGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZCCHC2 PubMed Links

Gene Details

Gene
ZCCHC2
Gene Name
zinc finger CCHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017742.5 548 Silent Mutation GAA,GAG E257E NP_060212.4
XM_006722493.3 548 Silent Mutation GAA,GAG E177E XP_006722556.1
XM_011526044.1 548 Intron XP_011524346.1
XM_017025802.1 548 Silent Mutation GAA,GAG E177E XP_016881291.1

View Full Product Details