Product Details
- SNP ID
-
rs3746634
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:3689414 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCCAGGGCAGAGAGGAGGAAAGAA[A/G]GAAACTACAGCAAGGAGGATTTAGG
- Phenotype
-
MIM: 607114
MIM: 600751
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139014307] are located under a probe and SNP(s) [rs16988862] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAM33
- Gene Name
- ADAM metallopeptidase domain 33
There are no transcripts associated with this gene.
- Gene
- SIGLEC1
- Gene Name
- sialic acid binding Ig like lectin 1
View Full Product Details