Product Details

SNP ID
rs113832750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107872603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCCCTAAAGTAATTTATATTC[A/T]ATGTTCTAAACTGTTACACTCCTTA
Phenotype
MIM: 608648
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 Intron NP_009145.1
XM_011535399.2 Intron XP_011533701.1
XM_017010218.1 Intron XP_016865707.1

View Full Product Details