Product Details

SNP ID
rs61747598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAATAATGGCGTAAGTCAGATTC[A/G]CACTTTCGATCTTCCTGCACTGTCA
Phenotype
MIM: 123910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 569 Silent Mutation TGC,TGT C173C NP_004122.2
XM_011536685.2 569 Silent Mutation TGC,TGT C161C XP_011534987.1

View Full Product Details