Product Details

SNP ID
rs61750008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143443582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTCATCACTGCTGCTCTGGGC[A/G]TGGAGTGGGTGCTACGGAGAATGTT
Phenotype
MIM: 613968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA1-AS1 PubMed Links

Gene Details

Gene
EPHA1-AS1
Gene Name
EPHA1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAS2R60
Gene Name
taste 2 receptor member 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177437.1 130 Missense Mutation ATG,GTG M44V NP_803186.1

View Full Product Details