Product Details

SNP ID
rs61750030
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:247605915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTCTCTGGCACTGGGCTCCAC[G/T]GAATGTATCCTCTTGGCTGACATGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2G3 PubMed Links

Gene Details

Gene
OR2G3
Gene Name
olfactory receptor family 2 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001914.1 330 Silent Mutation ACG,ACT T110T NP_001001914.1

View Full Product Details