Product Details

SNP ID
rs34974550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102766542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCAATCCAGTGGATTTTATG[A/G]GGAGCTTGCATTTTCTGTGCCACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
There are no transcripts associated with this gene.

Gene
TEX30
Gene Name
testis expressed 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286775.1 797 Missense Mutation NP_001273704.1
NM_001286776.1 797 Silent Mutation NP_001273705.1
NM_138779.4 797 Silent Mutation NP_620134.3
XM_005254097.2 797 Silent Mutation XP_005254154.1
XM_011521135.2 797 Silent Mutation XP_011519437.1
XM_011521136.2 797 Silent Mutation XP_011519438.1
XM_017020852.1 797 Silent Mutation XP_016876341.1

View Full Product Details