Product Details

SNP ID
rs112102379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:85678720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCACCCGCCCAGACGCTCGCGAA[A/G]GCCGACGTAAAAGACCGAAGTGCGC
Phenotype
MIM: 613263 MIM: 613117 MIM: 613264
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNHG5 PubMed Links

Gene Details

Gene
SNHG5
Gene Name
small nucleolar RNA host gene 5
There are no transcripts associated with this gene.

Gene
SNORD50A
Gene Name
small nucleolar RNA, C/D box 50A
There are no transcripts associated with this gene.

Gene
SNORD50B
Gene Name
small nucleolar RNA, C/D box 50B
There are no transcripts associated with this gene.

View Full Product Details