Product Details

SNP ID
rs185283820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:15595316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTGAAAATCAGGTTTTTTTTG[C/T]GATTTTCTAAATTATGGAAAGTTTC
Phenotype
MIM: 601257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX1 PubMed Links

Gene Details

Gene
DDX1
Gene Name
DEAD/H-box helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004939.2 Intron NP_004930.1

View Full Product Details