Product Details

SNP ID
rs3749394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131363371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTTACAGGTGCTGGGCCTGGTG[C/T]GAGTGCCCCTGTACCCTGCATGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC339874 PubMed Links

Gene Details

Gene
LOC339874
Gene Name
uncharacterized LOC339874
There are no transcripts associated with this gene.

Gene
NUDT16P1
Gene Name
nudix hydrolase 16 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details