Product Details

SNP ID
rs115922965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102766143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAATTCAGATGTTTAATTTTTT[C/T]CAAAACTTAACTATACTAATGAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
There are no transcripts associated with this gene.

Gene
TEX30
Gene Name
testis expressed 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286775.1 1196 UTR 3 NP_001273704.1
NM_001286776.1 1196 UTR 3 NP_001273705.1
NM_138779.4 1196 UTR 3 NP_620134.3
XM_005254097.2 1196 UTR 3 XP_005254154.1
XM_011521135.2 1196 UTR 3 XP_011519437.1
XM_011521136.2 1196 UTR 3 XP_011519438.1
XM_017020852.1 1196 UTR 3 XP_016876341.1

View Full Product Details