Product Details

SNP ID
rs151007769
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57682060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCAGTTTCTGCAGTGGAGGTCG[C/T]GACTGAGGGACGGGACAGAGAAGTC
Phenotype
MIM: 613419
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF551 PubMed Links
Additional Information
For this assay, SNP(s) [rs145547032] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ZNF551
Gene Name
zinc finger protein 551
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270938.1 92 Intron NP_001257867.1
NM_138347.4 92 UTR 5 NP_612356.2
Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
There are no transcripts associated with this gene.

View Full Product Details