Product Details

SNP ID
rs17049389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:58161929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGGTGACTATATTGCTGTATTA[C/T]AGGAAATATAATTACATTACTATTT
Phenotype
MIM: 608111 MIM: 602169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FANCL PubMed Links

Gene Details

Gene
FANCL
Gene Name
Fanconi anemia complementation group L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114636.1 4600 Intron NP_001108108.1
NM_018062.3 4600 Intron NP_060532.2
XM_005264395.3 4600 Intron XP_005264452.1
XM_005264397.4 4600 Intron XP_005264454.1
XM_011532939.2 4600 Intron XP_011531241.1
XM_011532940.2 4600 Intron XP_011531242.1
XM_011532941.1 4600 Intron XP_011531243.1
XM_011532942.2 4600 Intron XP_011531244.1
XM_011532944.2 4600 Intron XP_011531246.1
XM_011532945.2 4600 Intron XP_011531247.1
XM_017004414.1 4600 Intron XP_016859903.1
XM_017004415.1 4600 Intron XP_016859904.1
XM_017004416.1 4600 Intron XP_016859905.1
Gene
VRK2
Gene Name
vaccinia related kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130480.2 4600 Intron NP_001123952.1
NM_001130481.2 4600 Intron NP_001123953.1
NM_001130482.2 4600 Intron NP_001123954.1
NM_001130483.2 4600 Intron NP_001123955.1
NM_001288836.1 4600 Intron NP_001275765.1
NM_001288837.1 4600 Intron NP_001275766.1
NM_001288838.1 4600 Intron NP_001275767.1
NM_001288839.1 4600 Intron NP_001275768.1
NM_006296.6 4600 Intron NP_006287.2
XM_005264540.4 4600 Intron XP_005264597.1
XM_006712090.3 4600 Intron XP_006712153.1
XM_006712091.3 4600 UTR 3 XP_006712154.1
XM_006712092.3 4600 UTR 3 XP_006712155.1
XM_006712093.3 4600 UTR 3 XP_006712156.1
XM_011533092.2 4600 Intron XP_011531394.1
XM_017004858.1 4600 Intron XP_016860347.1
XM_017004859.1 4600 UTR 3 XP_016860348.1
XM_017004860.1 4600 UTR 3 XP_016860349.1
XM_017004861.1 4600 UTR 3 XP_016860350.1
XM_017004862.1 4600 UTR 3 XP_016860351.1
XM_017004863.1 4600 UTR 3 XP_016860352.1
XM_017004864.1 4600 UTR 3 XP_016860353.1
XM_017004865.1 4600 UTR 3 XP_016860354.1
XM_017004866.1 4600 UTR 3 XP_016860355.1

View Full Product Details