Product Details

SNP ID
rs61734768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104802364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTAGTGCACACTCGAGAGAAGCC[A/G]CATGCCTGCCGGTGGTGTGAGCGCC
Phenotype
MIM: 164730 MIM: 613915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKT1 PubMed Links

Gene Details

Gene
AKT1
Gene Name
AKT serine/threonine kinase 1
There are no transcripts associated with this gene.

Gene
ZBTB42
Gene Name
zinc finger and BTB domain containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137601.2 1184 Silent Mutation CCA,CCG P389P NP_001131073.1
XM_017020911.1 1184 Silent Mutation CCA,CCG P389P XP_016876400.1

View Full Product Details