Product Details

SNP ID
rs957400301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37084655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGGCCATGACTACCGTATCAC[A/G]CCGTCAGCCACTTTTCCATCCGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH11 PubMed Links

Gene Details

Gene
GPATCH11
Gene Name
G-patch domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278505.1 Intron NP_001265434.1
NM_001322249.1 Intron NP_001309178.1
NM_174931.3 Intron NP_777591.3
Gene
HEATR5B
Gene Name
HEAT repeat containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019024.2 Intron NP_061897.1

View Full Product Details