Product Details

SNP ID
rs16840274
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:158491415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTCTTTCTAGGTATTCTTTCTA[C/T]CTCTGAAATCTTCTACACAACTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10R3P PubMed Links

Gene Details

Gene
OR10R3P
Gene Name
olfactory receptor family 10 subfamily R member 3 pseudogene
There are no transcripts associated with this gene.

View Full Product Details