Product Details

SNP ID
rs61735869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168876112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTAGTCCAAGTCGATCTTTATA[C/G]AAGTCTGCAGATTTCTGCAGCCTTT
Phenotype
MIM: 609395
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPC25 PubMed Links

Gene Details

Gene
SPC25
Gene Name
SPC25, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020675.3 551 Missense Mutation TTC,TTG F137L NP_065726.1
XM_011511516.2 551 Missense Mutation TTC,TTG F137L XP_011509818.1

View Full Product Details