Product Details

SNP ID
rs7289828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30525262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTGTGCCCACCAGAACCAAGG[G/T]CCCTCACCTGGTAGGACCTGGATGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC14L6 PubMed Links

Gene Details

Gene
SEC14L6
Gene Name
SEC14 like lipid binding 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193336.2 Intron NP_001180265.2
XM_011530357.2 Intron XP_011528659.1
XM_011530358.2 Intron XP_011528660.1
XM_011530360.2 Intron XP_011528662.1
XM_017028931.1 Intron XP_016884420.1
XM_017028932.1 Intron XP_016884421.1
XM_017028933.1 Intron XP_016884422.1
XM_017028934.1 Intron XP_016884423.1

View Full Product Details