Product Details

SNP ID
rs545401987
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19721971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAATAATTTTAGTGGATGCTC[A/C]GGAGAGATAAATGCAAATGCAAAAA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TMEM196 PubMed Links
Additional Information
For this assay, SNP(s) [rs200443018] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TMEM196
Gene Name
transmembrane protein 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152774.3 1645 UTR 3 NP_689987.3
XM_005249692.4 1645 UTR 3 XP_005249749.2
XM_011515249.2 1645 UTR 3 XP_011513551.1
XM_011515250.2 1645 UTR 3 XP_011513552.1
XM_017011928.1 1645 Intron XP_016867417.1
XM_017011929.1 1645 Intron XP_016867418.1

View Full Product Details