Product Details

SNP ID
rs150845780
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:53216677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACAATTATTACTACCGCTAGAA[A/G]GGAGAATGTCTAGTCCATGGTATCT
Phenotype
MIM: 600650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf123 PubMed Links

Gene Details

Gene
C1orf123
Gene Name
chromosome 1 open reading frame 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304759.1 Intron NP_001291688.1
NM_001304760.1 Intron NP_001291689.1
NM_017887.2 Intron NP_060357.1
Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details