Product Details

SNP ID
rs34377647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56995839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCTTCATGGCACCTTTTGAAAC[A/G]TACAGCACCTGGGCCCTGGCGGTGG
Phenotype
MIM: 605307
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007264.3 1400 Silent Mutation ACA,ACG T210T NP_009195.1
XM_011537824.2 1400 Silent Mutation ACA,ACG T210T XP_011536126.1
XM_011537825.2 1400 Silent Mutation ACA,ACG T210T XP_011536127.1
XM_011537827.2 1400 Silent Mutation ACA,ACG T210T XP_011536129.1
Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details