Product Details

SNP ID
hCV25936272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19717988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGGGCCGGCTCTGAGGCGGGA[A/C]AACTTTCTGGAGGCGGCGGCCTCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IQCK PubMed Links

Gene Details

Gene
IQCK
Gene Name
IQ motif containing K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305121.1 381 UTR 5 NP_001292050.1
NM_001305122.1 381 UTR 5 NP_001292051.1
NM_153208.2 381 UTR 5 NP_694940.1
Gene
KNOP1
Gene Name
lysine rich nucleolar protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012991.2 381 Intron NP_001013009.2

View Full Product Details