Product Details

SNP ID
rs3748556
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:196957826 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GATAGACTATAAAGTGCCTTGTTTG[C/T]ATTTGCCTTATTTGAACTTGTATTT
Phenotype
MIM: 600889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFHR2 PubMed Links

Gene Details

Gene
CFHR2
Gene Name
complement factor H related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312672.1 Intron NP_001299601.1
NM_005666.3 Intron NP_005657.1
XM_005245113.3 Intron XP_005245170.1
XM_011509458.2 Intron XP_011507760.1
XM_011509459.2 Intron XP_011507761.1
XM_011509460.2 Intron XP_011507762.1
XM_017001109.1 Intron XP_016856598.1

View Full Product Details