Product Details

SNP ID
rs34656572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127878256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGATGATGGTTACAGTGACTA[C/T]ATTTAGAAGAATGACGAGCAGTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLRN3 PubMed Links

Gene Details

Gene
CLRN3
Gene Name
clarin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152311.3 556 Missense Mutation ATA,GTA I192V NP_689524.1
XM_011539274.2 556 Missense Mutation ATA,GTA I132V XP_011537576.1

View Full Product Details