Product Details

SNP ID
rs34392164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:281405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGGCCTCAGGAGGAGGAGTGCG[A/G]GTATCCATAGTGGTTGTGGTCAGCC
Phenotype
MIM: 607126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLPP2 PubMed Links

Gene Details

Gene
PLPP2
Gene Name
phospholipid phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003712.3 949 Missense Mutation CCG,TCG P284S NP_003703.1
NM_177526.2 949 Missense Mutation CCG,TCG P228S NP_803545.1
NM_177543.2 949 Missense Mutation CCG,TCG P305S NP_808211.1
XM_011528396.2 949 Missense Mutation CCG,TCG P290S XP_011526698.1

View Full Product Details