Product Details

SNP ID
rs35862579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33722972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTTCCTGCCCATCATAGATGAC[A/G]AGGAGAGAGCTGGAATAGAAGCGGT
Phenotype
MIM: 606993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IP6K3 PubMed Links

Gene Details

Gene
IP6K3
Gene Name
inositol hexakisphosphate kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142883.1 1408 Silent Mutation CTC,CTT L327L NP_001136355.1
NM_054111.4 1408 Silent Mutation CTC,CTT L327L NP_473452.2
XM_005248842.3 1408 Silent Mutation CTC,CTT L327L XP_005248899.1
XM_005248843.3 1408 Silent Mutation CTC,CTT L214L XP_005248900.1
XM_011514295.2 1408 Intron XP_011512597.1

View Full Product Details