Product Details

SNP ID
rs116357503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73860790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTTATATATTGATACCTTTTA[A/G]TTATAGATTGCCTTTGTTTTTATTC
Phenotype
MIM: 608642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGR2 PubMed Links

Gene Details

Gene
PTGR2
Gene Name
prostaglandin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146154.1 Intron NP_001139626.1
NM_001146155.1 Intron NP_001139627.1
NM_152444.2 Intron NP_689657.1

View Full Product Details