Product Details

SNP ID
rs773383521
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:197912506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCAGTGCAACCACCATTACGG[A/C]GTGATCCACTCCTTTTCCTCTAAGA
Phenotype
MIM: 606066
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf53 PubMed Links

Gene Details

Gene
C1orf53
Gene Name
chromosome 1 open reading frame 53
There are no transcripts associated with this gene.

Gene
LHX9
Gene Name
LIM homeobox 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014434.1 2 UTR 5 NP_001014434.1
NM_020204.2 2 Intron NP_064589.2
XM_005245350.3 2 Intron XP_005245407.2
XM_011509781.2 2 Intron XP_011508083.2
XM_017001849.1 2 Intron XP_016857338.1

View Full Product Details