Product Details

SNP ID
rs116572693
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:20164182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACAGAGACACAGAGGGTCACTG[A/G]GGGCTCCCAGCCCAGCCCCAGGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2C PubMed Links

Gene Details

Gene
PLA2G2C
Gene Name
phospholipase A2 group IIC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316722.1 Intron NP_001303651.1
XM_017001270.1 Intron XP_016856759.1

View Full Product Details