Product Details

SNP ID
rs200936716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128060698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTCCCCTGGTGGCGACAGCTTT[C/T]AGCAAAAACAATTGAGCAATGCAAA
Phenotype
MIM: 603449 MIM: 609213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RUVBL1 PubMed Links

Gene Details

Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
There are no transcripts associated with this gene.

Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 Intron NP_037468.1

View Full Product Details