Product Details

SNP ID
hCV25943502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41268406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAATCTACATGGAGCCTTTACC[C/T]CCTGCTTAGAGCGGGTCTTCCCAGA
Phenotype
MIM: 109135 MIM: 605800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
HNRNPUL1
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301016.2 Intron NP_001287945.1
NM_001321208.1 Intron NP_001308137.1
NM_001321211.1 Intron NP_001308140.1
NM_007040.5 Intron NP_008971.2
NM_144732.4 Intron NP_653333.1

View Full Product Details