Product Details
- SNP ID
-
rs61739185
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:95315104 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGTTGCCTGGAAAAGTACCAAA[C/T]TGCTTTGTCCTTCTTGAAGTACCTA
- Phenotype
-
MIM: 605264
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SORBS1
PubMed Links
Gene Details
- Gene
- SORBS1
- Gene Name
- sorbin and SH3 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001034954.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1278Q |
NP_001030126.1 |
NM_001034955.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1137Q |
NP_001030127.1 |
NM_001034956.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q891Q |
NP_001030128.1 |
NM_001034957.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q726Q |
NP_001030129.1 |
NM_001290294.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1252Q |
NP_001277223.1 |
NM_001290295.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q735Q |
NP_001277224.1 |
NM_001290296.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q990Q |
NP_001277225.1 |
NM_001290297.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q797Q |
NP_001277226.1 |
NM_001290298.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q832Q |
NP_001277227.1 |
NM_006434.2 |
3572 |
Silent Mutation |
CAA,CAG |
Q670Q |
NP_006425.2 |
NM_015385.3 |
3572 |
Silent Mutation |
CAA,CAG |
Q802Q |
NP_056200.1 |
NM_024991.2 |
3572 |
Silent Mutation |
CAA,CAG |
Q767Q |
NP_079267.1 |
XM_006717589.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q990Q |
XP_006717652.1 |
XM_006717593.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q726Q |
XP_006717656.1 |
XM_011539140.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1516Q |
XP_011537442.1 |
XM_011539150.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1491Q |
XP_011537452.1 |
XM_011539155.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1366Q |
XP_011537457.1 |
XM_011539167.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1127Q |
XP_011537469.1 |
XM_017015500.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1351Q |
XP_016870989.1 |
XM_017015501.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1213Q |
XP_016870990.1 |
XM_017015502.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1182Q |
XP_016870991.1 |
XM_017015503.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1157Q |
XP_016870992.1 |
XM_017015504.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1154Q |
XP_016870993.1 |
XM_017015505.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1129Q |
XP_016870994.1 |
XM_017015506.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1129Q |
XP_016870995.1 |
XM_017015507.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1126Q |
XP_016870996.1 |
XM_017015508.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1098Q |
XP_016870997.1 |
XM_017015509.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1097Q |
XP_016870998.1 |
XM_017015510.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1073Q |
XP_016870999.1 |
XM_017015511.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1070Q |
XP_016871000.1 |
XM_017015512.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1032Q |
XP_016871001.1 |
XM_017015513.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1029Q |
XP_016871002.1 |
XM_017015514.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q1001Q |
XP_016871003.1 |
XM_017015515.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q943Q |
XP_016871004.1 |
XM_017015516.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q934Q |
XP_016871005.1 |
XM_017015517.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q915Q |
XP_016871006.1 |
XM_017015518.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q911Q |
XP_016871007.1 |
XM_017015519.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q902Q |
XP_016871008.1 |
XM_017015520.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q890Q |
XP_016871009.1 |
XM_017015521.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q886Q |
XP_016871010.1 |
XM_017015522.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q881Q |
XP_016871011.1 |
XM_017015523.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q858Q |
XP_016871012.1 |
XM_017015524.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q849Q |
XP_016871013.1 |
XM_017015525.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q844Q |
XP_016871014.1 |
XM_017015526.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q820Q |
XP_016871015.1 |
XM_017015527.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q819Q |
XP_016871016.1 |
XM_017015528.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q811Q |
XP_016871017.1 |
XM_017015529.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q795Q |
XP_016871018.1 |
XM_017015530.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q792Q |
XP_016871019.1 |
XM_017015531.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q788Q |
XP_016871020.1 |
XM_017015532.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q786Q |
XP_016871021.1 |
XM_017015533.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q779Q |
XP_016871022.1 |
XM_017015534.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q764Q |
XP_016871023.1 |
XM_017015535.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q763Q |
XP_016871024.1 |
XM_017015536.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q760Q |
XP_016871025.1 |
XM_017015537.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q758Q |
XP_016871026.1 |
XM_017015538.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q755Q |
XP_016871027.1 |
XM_017015539.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q754Q |
XP_016871028.1 |
XM_017015540.1 |
3572 |
Silent Mutation |
CAA,CAG |
Q751Q |
XP_016871029.1 |
View Full Product Details