Product Details
- SNP ID
-
rs112315831
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:232984520 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAACAGTGAACAGCTCAGCAAACG[A/T]TCACTACCCACTTCTAACTGGCATC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NTPCR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113070999] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NTPCR
- Gene Name
- nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.
- Gene
- PCNX2
- Gene Name
- pecanex homolog 2 (Drosophila)
View Full Product Details