Product Details

SNP ID
rs34553814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12310965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAGAAATATGTGCCCAGGGC[C/T]GCCCTGGTGGACTTAGAGCCAGGCA
Phenotype
MIM: 615103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBB6 PubMed Links

Gene Details

Gene
TUBB6
Gene Name
tubulin beta 6 class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303524.1 423 Silent Mutation GCC,GCT A63A NP_001290453.1
NM_001303525.1 423 Silent Mutation GCC,GCT A63A NP_001290454.1
NM_001303526.1 423 Intron NP_001290455.1
NM_001303527.1 423 UTR 5 NP_001290456.1
NM_001303528.1 423 UTR 5 NP_001290457.1
NM_001303529.1 423 UTR 5 NP_001290458.1
NM_001303530.1 423 UTR 5 NP_001290459.1
NM_032525.2 423 Silent Mutation GCC,GCT A63A NP_115914.1

View Full Product Details