Product Details

SNP ID
rs4657093
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:161723213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTGGTAGTCTGTAACCCAATGAA[C/T]GTGATCTCCTCTCCTGCAGGGATGG
Phenotype
MIM: 606891 MIM: 609251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCRLA PubMed Links

Gene Details

Gene
FCRLA
Gene Name
Fc receptor like A
There are no transcripts associated with this gene.

Gene
FCRLB
Gene Name
Fc receptor like B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002901.3 Intron NP_001002901.1
NM_001288829.1 Intron NP_001275758.1
NM_001288830.1 Intron NP_001275759.1
NM_001288831.1 Intron NP_001275760.1
NM_001288832.1 Intron NP_001275761.1
NM_001320241.1 Intron NP_001307170.1

View Full Product Details