Product Details

SNP ID
hCV25953404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35809129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTTCTTTGATTCCTCATTCCA[C/T]CATCCTCCCCATTCCACCCACCCCA
Phenotype
MIM: 609997 MIM: 108961 MIM: 605731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM221B PubMed Links

Gene Details

Gene
FAM221B
Gene Name
family with sequence similarity 221 member B
There are no transcripts associated with this gene.

Gene
HINT2
Gene Name
histidine triad nucleotide binding protein 2
There are no transcripts associated with this gene.

Gene
NPR2
Gene Name
natriuretic peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003995.3 Intron NP_003986.2
Gene
SPAG8
Gene Name
sperm associated antigen 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039592.1 Intron NP_001034681.1
NM_172312.1 Intron NP_758516.1

View Full Product Details