Product Details
- SNP ID
-
rs3735326
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149839687 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAATTTCAAAAGAGGTGATTTCT[C/G]GCATGAGCATCTCTTCCATCCTCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SSPO
PubMed Links
Gene Details
- Gene
- SSPO
- Gene Name
- SCO-spondin
There are no transcripts associated with this gene.
- Gene
- ZNF862
- Gene Name
- zinc finger protein 862
View Full Product Details