Product Details

SNP ID
rs4764506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6652860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTTTCCAACCTGGTGCCAAACT[C/T]CCCACCAGATGAAGAGCTGACTTCT
Phenotype
MIM: 608352 MIM: 608524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACRBP PubMed Links

Gene Details

Gene
ACRBP
Gene Name
acrosin binding protein
There are no transcripts associated with this gene.

Gene
ING4
Gene Name
inhibitor of growth family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127582.1 Intron NP_001121054.1
NM_001127583.1 Intron NP_001121055.1
NM_001127584.1 Intron NP_001121056.1
NM_001127585.1 Intron NP_001121057.1
NM_001127586.1 Intron NP_001121058.1
NM_016162.3 Intron NP_057246.2
XM_005253698.3 Intron XP_005253755.1
XM_011520964.2 Intron XP_011519266.1
XM_011520965.2 Intron XP_011519267.1
XM_017019392.1 Intron XP_016874881.1
XM_017019393.1 Intron XP_016874882.1

View Full Product Details