Product Details

SNP ID
rs115150402
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:32180263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGTGGGGATTTCTGATCCAGG[C/T]TGCGAAGAATTTCGAAGTCTGGAAA
Phenotype
MIM: 610563 MIM: 608676
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KPNA6 PubMed Links

Gene Details

Gene
KPNA6
Gene Name
karyopherin subunit alpha 6
There are no transcripts associated with this gene.

Gene
TXLNA
Gene Name
taxilin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175852.3 276 Intron NP_787048.1
XM_011540931.2 276 Intron XP_011539233.1
XM_011540932.2 276 Intron XP_011539234.1
XM_017000561.1 276 UTR 5 XP_016856050.1
XM_017000562.1 276 UTR 5 XP_016856051.1
XM_017000563.1 276 Intron XP_016856052.1
XM_017000564.1 276 Intron XP_016856053.1
XM_017000565.1 276 Intron XP_016856054.1

View Full Product Details