Product Details

SNP ID
rs34988750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20189104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCAGCCGCTGCATGGAGAAGC[C/T]GTCGTACATGACATAGAAGACGTAC
Phenotype
MIM: 121011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJB2 PubMed Links

Gene Details

Gene
GJB2
Gene Name
gap junction protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004004.5 684 Missense Mutation AGC,GGC S160G NP_003995.2
XM_011535049.2 684 Missense Mutation AGC,GGC S160G XP_011533351.1

View Full Product Details