Product Details

SNP ID
rs148840229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13738595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAAGGCATTAAACATTTAGGCA[C/T]ATCATCTTTGATGTGAAGTACTTAG
Phenotype
MIM: 300170 MIM: 300202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OFD1 PubMed Links

Gene Details

Gene
OFD1
Gene Name
OFD1, centriole and centriolar satellite protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003611.2 Intron NP_003602.1
XM_005274604.2 Intron XP_005274661.1
XM_005274606.3 Intron XP_005274663.1
XM_011545592.2 Intron XP_011543894.1
XM_011545594.2 Intron XP_011543896.1
XM_011545595.1 Intron XP_011543897.1
XM_011545597.1 Intron XP_011543899.1
XM_011545598.1 Intron XP_011543900.1
XM_017029909.1 Intron XP_016885398.1
XM_017029910.1 Intron XP_016885399.1
XM_017029911.1 Intron XP_016885400.1
Gene
TRAPPC2
Gene Name
trafficking protein particle complex 2
There are no transcripts associated with this gene.

View Full Product Details