Product Details

SNP ID
rs34480825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47838400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCAAAAGTGGGGGACAGCACC[C/T]GGGGGGCCTGGGCCACCCCCATCCC
Phenotype
MIM: 611825 MIM: 614966 MIM: 608613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC46 PubMed Links

Gene Details

Gene
LRRC46
Gene Name
leucine rich repeat containing 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033413.3 1043 Intron NP_219481.1
XM_005257775.4 1043 Intron XP_005257832.1
Gene
MRPL10
Gene Name
mitochondrial ribosomal protein L10
There are no transcripts associated with this gene.

Gene
SCRN2
Gene Name
secernin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145023.1 1043 Missense Mutation CAG,CGG Q330R NP_001138495.1
NM_138355.3 1043 Missense Mutation CAG,CGG Q330R NP_612364.2
XM_005257776.4 1043 Missense Mutation CAG,CGG Q330R XP_005257833.1
XM_017025286.1 1043 Missense Mutation CAG,CGG Q330R XP_016880775.1
Gene
SP6
Gene Name
Sp6 transcription factor
There are no transcripts associated with this gene.

View Full Product Details