Product Details

SNP ID
rs17832885
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58207751 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCACAGACTAAAACATTCTGCCAT[C/T]TTAGACATTCCGATTCTTAAGGATT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACTR10 PubMed Links

Gene Details

Gene
ACTR10
Gene Name
actin-related protein 10 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018477.2 Intron NP_060947.1
XM_011536960.1 Intron XP_011535262.1
XM_011536961.1 Intron XP_011535263.1
XM_011536962.2 Intron XP_011535264.1
XM_011536963.2 Intron XP_011535265.1

View Full Product Details