Product Details
- SNP ID
-
hCV25958535
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:38110701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCCCTTGGTAGTAGAGATGTAGT[A/T]CTACAGATAACTGTTTCTTCCTTTT
- Phenotype
-
MIM: 604782
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ASH2L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs376065614] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASH2L
- Gene Name
- ASH2 like histone lysine methyltransferase complex subunit
View Full Product Details