Product Details

SNP ID
rs7520406
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:16006004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCACCCACGGGAAGCAGCTCTC[C/T]CTGGCAGCAACCGCATCACCACCCC
Phenotype
MIM: 610692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf64 PubMed Links

Gene Details

Gene
C1orf64
Gene Name
chromosome 1 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178840.3 236 Silent Mutation TCC,TCT S56S NP_849162.1
Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
There are no transcripts associated with this gene.

View Full Product Details